Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699335.3 | XP_005699392.3 | 2 - 3 | 1657 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |