Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962503.2 | XP_013817957.2 | 1 - 2 | 160 | 86 | |
XM_018045958.1 | XP_017901447.1 | 1 - 2 | 160 | 86 | ||
XM_018045959.1 | XP_017901448.1 | 1 - 2 | 160 | 86 | ||
XM_018045961.1 | XP_017901450.1 | 1 - 2 | 160 | 86 | ||
XM_018045962.1 | XP_017901451.1 | 1 - 2 | 160 | 86 | ||
GenBank mRNA | JO559054.2 | - | 1 - 2 | 172 | 86 | |
JO562932.2 | - | 1 - 2 | 172 | 86 | ||
JO586094.2 | - | 5 - 6 | 2270 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |