Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962503.2 | XP_013817957.2 | 3 - 4 | 732 | 74 | |
XM_018045958.1 | XP_017901447.1 | 3 - 4 | 732 | 74 | ||
XM_018045959.1 | XP_017901448.1 | 3 - 4 | 732 | 74 | ||
XM_018045960.1 | XP_017901449.1 | 3 - 4 | 977 | 74 | ||
GenBank mRNA | JO559054.2 | - | 3 - 4 | 744 | 74 | |
JO562932.2 | - | 3 - 4 | 744 | 74 | ||
JO586094.2 | - | 3 - 4 | 1698 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |