Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102188370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677751.2 | XP_005677808.1 | 8 - 9 | 880 | 98 | |
XM_005677753.3 | XP_005677810.1 | 8 - 9 | 881 | 98 | ||
XM_018046005.1 | XP_017901494.1 | 6 - 7 | 581 | 98 | ||
GenBank mRNA | JO594747.2 | - | 8 - 9 | 839 | 98 | |
JO646858.2 | - | 2 - 3 | 127 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Capra hircus Probe spans exons |