Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694594.3 | XP_005694651.2 | 7 - 8 | 1139 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |