Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047432.1 | XP_017902921.1 | 3 - 4 | 635 | 109 | |
XM_018047433.1 | XP_017902922.1 | 3 - 4 | 577 | 109 | ||
GenBank mRNA | JT665159.1 | - | 3 - 4 | 504 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |