Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043250.1 | XP_017898739.1 | 3 - 4 | 809 | 82 | |
XM_018043251.1 | XP_017898740.1 | 3 - 4 | 725 | 82 | ||
GenBank mRNA | JO534854.2 | - | 3 - 4 | 778 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |