Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061469.1 | XP_017916958.1 | 1 - 2 | 143 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |