Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055530.1 | XP_017911019.1 | 3 - 4 | 460 | 74 | |
XM_018055531.1 | XP_017911020.1 | 3 - 4 | 460 | 74 | ||
GenBank mRNA | KA306344.1 | - | 3 - 4 | 311 | 74 |
Gene Symbol: LOC108633924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041082.1 | XP_017896571.1 | 1 | 394 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |