Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066630.1 | XP_017922119.1 | 3 - 4 | 641 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |