Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050876.1 | XP_017906365.1 | 5 - 6 | 4833 | 88 | |
XM_018050877.1 | XP_017906366.1 | 6 - 7 | 4734 | 88 | ||
GenBank mRNA | JO362555.2 | - | 1 - 2 | 475 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |