Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041182.1 | XP_017896671.1 | 4 - 5 | 467 | 94 | |
XM_018041183.1 | XP_017896672.1 | 4 - 5 | 437 | 94 | ||
XM_018041188.1 | XP_017896677.1 | 2 - 3 | 206 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |