Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974043.2 | XP_013829497.1 | 4 - 5 | 2204 | 82 | |
XM_013974048.2 | XP_013829502.1 | 5 - 6 | 2145 | 82 | ||
XM_018056505.1 | XP_017911994.1 | 4 - 5 | 1403 | 82 | ||
XM_018056509.1 | XP_017911998.1 | 4 - 5 | 1374 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |