Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064067.1 | XP_017919556.1 | 3 - 4 | 1325 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |