Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061796.1 | XP_017917285.1 | 2 - 3 | 321 | 121 | |
XM_018061797.1 | XP_017917286.1 | 2 - 3 | 212 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |