Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678537.3 | XP_005678594.2 | 4 - 5 | 512 | 62 | |
XM_005678538.3 | XP_005678595.2 | 4 - 5 | 480 | 62 | ||
XM_018043596.1 | XP_017899085.1 | 4 - 5 | 512 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |