Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698620.3 | XP_005698677.2 | 1 - 2 | 983 | 82 | |
GenBank mRNA | KA334949.1 | - | 1 - 2 | 518 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |