Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709613.3 | XP_005709670.2 | 3 - 4 | 653 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |