Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681055.3 | XP_005681112.1 | 3 - 4 | 316 | 89 | |
XM_013964330.2 | XP_013819784.1 | 3 - 4 | 715 | 89 | ||
GenBank mRNA | JO422482.2 | - | 1 - 2 | 310 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |