Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682278.3 | XP_005682335.1 | 3 - 4 | 428 | 68 | |
XM_005682279.2 | XP_005682336.1 | 3 - 4 | 426 | 68 | ||
XM_005682280.3 | XP_005682337.1 | 3 - 4 | 440 | 68 | ||
XM_005682281.2 | XP_005682338.1 | 3 - 4 | 393 | 68 | ||
GenBank mRNA | JO588909.2 | - | 2 - 3 | 541 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |