Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060347.1 | XP_017915836.1 | 2 - 3 | 868 | 94 | |
GenBank mRNA | KA341751.1 | - | 2 - 3 | 192 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |