Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060347.1 | XP_017915836.1 | 3 - 4 | 1001 | 83 | |
GenBank mRNA | KA341751.1 | - | 1 - 2 | 59 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |