Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680398.3 | XP_005680455.1 | 4 - 5 | 1501 | 61 | |
XM_013963965.2 | XP_013819419.1 | 4 - 5 | 1596 | 61 | ||
GenBank mRNA | JO454228.2 | - | 1 - 2 | 338 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |