Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047025.1 | XP_017902514.1 | 4 - 5 | 858 | 56 | |
GenBank mRNA | JO611985.2 | - | 2 - 3 | 1930 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |