Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063759.1 | XP_017919248.1 | 2 - 3 | 856 | 78 | |
GenBank mRNA | JO626144.2 | - | 1 - 2 | 102 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |