Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973884.2 | XP_013829338.1 | 1 - 2 | 368 | 59 | |
XM_018039140.1 | XP_017894629.1 | 1 - 2 | 352 | 59 | ||
GenBank mRNA | JI861655.1 | - | 1 - 2 | 270 | 59 | |
JO644987.2 | - | 5 - 6 | 941 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |