Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973971.2 | XP_013829425.2 | 5 - 6 | 1097 | 98 | |
GenBank mRNA | KA314117.1 | - | 1 - 2 | 412 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |