Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047648.1 | XP_017903137.1 | 3 - 4 | 899 | 118 | |
XM_018047649.1 | XP_017903138.1 | 3 - 4 | 685 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |