Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054714.1 | XP_017910203.1 | 14 - 15 | 1196 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |