Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064747.1 | XP_017920236.1 | 4 - 5 | 459 | 104 | |
GenBank mRNA | JO630623.2 | - | 2 - 3 | 1292 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |