Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060981.1 | XP_017916470.1 | 3 - 4 | 575 | 102 | |
XM_018060982.1 | XP_017916471.1 | 3 - 4 | 572 | 102 | ||
GenBank mRNA | JO641020.2 | - | 3 - 4 | 532 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |