Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19H17orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064063.1 | XP_017919552.1 | 2 - 3 | 148 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |