Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694591.3 | XP_005694648.1 | 2 - 3 | 522 | 103 | |
GenBank mRNA | JO625361.2 | - | 2 - 3 | 283 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |