Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058007.1 | XP_017913496.1 | 2 - 3 | 1154 | 75 | |
GenBank mRNA | JO511990.2 | - | 1 - 2 | 596 | 75 | |
JO624692.2 | - | 1 - 2 | 597 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |