Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700197.3 | XP_005700254.2 | 3 | 956 | 179 | |
XM_018044278.1 | XP_017899767.1 | 3 | 969 | 179 | ||
GenBank mRNA | JO437002.2 | - | 1 | 234 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |