Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172969.1 | NP_001166440.1 | 3 - 4 | 414 | 109 | |
GenBank mRNA | AB058509.1 | - | 3 - 4 | 414 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |