Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172971.1 | NP_001166442.1 | 8 - 9 | 908 | 59 | |
GenBank mRNA | AF256520.1 | - | 8 - 9 | 908 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Cavia porcellus Probe spans exons |