Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172974.1 | NP_001166445.1 | 2 | 1450 | 98 | |
GenBank mRNA | AF183919.1 | - | 2 | 1450 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Cavia porcellus Both primers and probe map within a single exon |