Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172932.1 | NP_001166403.1 | 1 - 2 | 87 | 98 | |
GenBank mRNA | AM851020.1 | - | 1 - 2 | 87 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |