Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172932.1 | NP_001166403.1 | 2 - 3 | 237 | 73 | |
GenBank mRNA | AM851020.1 | - | 2 - 3 | 237 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Cavia porcellus Probe spans exons |