Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173109.1 | NP_001166580.1 | 1 - 2 | 154 | 95 | |
GenBank mRNA | AB010634.1 | - | 1 - 2 | 154 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |