Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172871.2 | NP_001166342.1 | 5 - 6 | 504 | 84 | |
GenBank mRNA | AJ306650.1 | - | 5 - 6 | 505 | 84 | |
EB369382.1 | - | 5 - 6 | 495 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |