Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173119.1 | NP_001166590.1 | 4 - 5 | 1260 | 88 | |
GenBank mRNA | U50545.1 | - | 4 - 5 | 1260 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Cavia porcellus Probe spans exons |