Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173074.1 | NP_001166545.1 | 1 - 2 | 514 | 88 | |
XM_005006539.2 | XP_005006596.1 | 1 - 2 | 496 | 88 | ||
GenBank mRNA | EU257503.1 | - | 1 - 2 | 514 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |