Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173074.1 | NP_001166545.1 | 3 - 4 | 750 | 66 | |
XM_005006539.2 | XP_005006596.1 | 3 - 4 | 732 | 66 | ||
GenBank mRNA | EU257503.1 | - | 3 - 4 | 750 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Cavia porcellus Probe spans exons |