Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172763.1 | NP_001166234.1 | 2 - 3 | 712 | 69 | |
GenBank mRNA | AY075097.1 | - | 2 - 3 | 712 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |