Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ku80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135923.3 | NP_609767.2 | 3 - 4 | 430 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |