Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkaap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135939.3 | NP_609783.1 | 2 - 3 | 456 | 65 | |
GenBank mRNA | BT050448.1 | - | 2 - 3 | 454 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |