Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIIc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135986.3 | NP_609830.2 | 6 - 7 | 1243 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |