Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pzg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275214.1 | NP_001262143.1 | 1 - 2 | 971 | 75 | |
NM_141040.2 | NP_649297.1 | 1 - 2 | 971 | 75 | ||
GenBank mRNA | BT010057.1 | - | 1 - 2 | 1007 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |